Assay Design



Gene Transcripts

Gene Symbol
pqn-62
Entrez Gene ID
180789
Gene Name
protein-coding
Gene Aliases
CELE_T03G11.1
Chromosome Location
Chr.X: 5196110 - 5204274 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313445.3 NP_001300374.1 13 - 14 2685 67
NM_001313446.4 NP_001300375.1 15 - 16 2982 67
NM_001380960.1 NP_001368582.1 15 - 16 2961 67

View Full Product Details