Assay Design
Gene Transcripts
- Gene Symbol
- ifta-1
- Entrez Gene ID
-
183817
- Gene Name
- WD_REPEATS_REGION domain-containing protein
- Gene Aliases
-
CELE_C54G7.4
- Chromosome Location
-
Chr.X: 5545587 - 5552756 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076539.6 |
NP_508940.4 |
22 - 23 |
3194 |
83 |
View Full Product Details