Assay Design



Gene Transcripts

Gene Symbol
egl-13
Entrez Gene ID
180833
Gene Name
protein-coding
Gene Aliases
CELE_T22B7.1
Chromosome Location
Chr.X: 5663835 - 5674067 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029747.6 NP_001024918.1 3 - 4 309 71
NM_001380981.1 NP_001368074.1 1 - 2 88 71
NR_138511.1 2 - 3 232 71
GenBank mRNA AF097319.1 - 2 - 3 240 71

View Full Product Details