Assay Design



Gene Transcripts

Gene Symbol
ctbp-1
Entrez Gene ID
180853
Gene Name
protein-coding
Gene Aliases
CELE_F49E10.5
Chromosome Location
Chr.X: 5862708 - 5873347 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270101.4 NP_001257030.1 6 - 7 691 57
NM_001270102.3 NP_001257031.1 3 - 4 328 57

View Full Product Details