Assay Design
Gene Transcripts
- Gene Symbol
- mom-1
- Entrez Gene ID
-
180904
- Gene Name
- Protein-serine O-palmitoleoyltransferase porcupine
- Gene Aliases
-
CELE_T07H6.2
- Chromosome Location
-
Chr.X: 6293124 - 6298305 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076653.5 |
NP_509054.1 |
2 - 3 |
346 |
120 |
GenBank mRNA |
AF013489.1 |
- |
2 - 3 |
343 |
120 |
View Full Product Details