Assay Design



Gene Transcripts

Gene Symbol
dop-3
Entrez Gene ID
188499
Gene Name
protein-coding
Gene Aliases
CELE_T14E8.3
Chromosome Location
Chr.X: 6564175 - 6569724 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029736.6 NP_001024907.2 10 - 11 1792 130
NM_001029737.5 NP_001024908.2 10 - 11 1596 130
NM_001038472.5 NP_001033561.1 9 - 10 1975 130
NM_001381012.1 NP_001368078.1 10 - 11 1558 130
GenBank mRNA AY485309.1 - 10 - 11 1592 130

View Full Product Details