Assay Design



Gene Transcripts

Gene Symbol
dop-3
Entrez Gene ID
188499
Gene Name
protein-coding
Gene Aliases
CELE_T14E8.3
Chromosome Location
Chr.X: 6564175 - 6569724 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029736.6 NP_001024907.2 3 - 4 716 88
NM_001381012.1 NP_001368078.1 3 - 4 482 88

View Full Product Details