Assay Design
Gene Transcripts
- Gene Symbol
- acr-3
- Entrez Gene ID
-
180937
- Gene Name
- Acetylcholine receptor subunit beta-type acr-3
- Gene Aliases
-
CELE_K11G12.7
- Chromosome Location
-
Chr.X: 6709538 - 6712925 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076728.4 |
NP_509129.1 |
5 - 6 |
612 |
78 |
GenBank mRNA |
Y08637.1 |
- |
- |
612 |
78 |
View Full Product Details