Assay Design
Gene Transcripts
- Gene Symbol
- atg-2
- Entrez Gene ID
-
180949
- Gene Name
- Autophagy-related protein 2
- Gene Aliases
-
CELE_M03A8.2
- Chromosome Location
-
Chr.X: 6806729 - 6817266 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076744.9 |
NP_509145.3 |
5 - 6 |
1055 |
73 |
View Full Product Details