Assay Design
Gene Transcripts
- Gene Symbol
- unc-6
- Entrez Gene ID
-
180961
- Gene Name
- Netrin unc-6
- Gene Aliases
-
CELE_F41C6.1
- Chromosome Location
-
Chr.X: 6889674 - 6897147 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076764.5 |
NP_509165.1 |
1 - 2 |
140 |
95 |
View Full Product Details