Assay Design
Gene Transcripts
- Gene Symbol
- otpl-3
- Entrez Gene ID
-
259710
- Gene Name
- OToPetrin-Like
- Gene Aliases
-
CELE_F14B8.7
- Chromosome Location
-
Chr.X: 6921646 - 6924764 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076769.4 |
NP_509170.1 |
7 - 8 |
919 |
134 |
View Full Product Details