Assay Design
Gene Transcripts
- Gene Symbol
- ubc-17
- Entrez Gene ID
-
181979
- Gene Name
- Probable ubiquitin-conjugating enzyme protein 17
- Gene Aliases
-
CELE_B0403.2
- Chromosome Location
-
Chr.X: 7050590 - 7055709 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076787.5 |
NP_509188.3 |
7 - 8 |
984 |
87 |
View Full Product Details