Assay Design
Gene Transcripts
- Gene Symbol
- lam-2
- Entrez Gene ID
-
180980
- Gene Name
- Laminin-like protein lam-2
- Gene Aliases
-
CELE_C54D1.5
- Chromosome Location
-
Chr.X: 7143846 - 7151239 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076803.7 |
NP_509204.3 |
5 - 6 |
1091 |
87 |
View Full Product Details