Assay Design



Gene Transcripts

Gene Symbol
glb-16
Entrez Gene ID
185848
Gene Name
GLoBin related
Gene Aliases
CELE_F46C8.7
Chromosome Location
Chr.X: 7537809 - 7540133 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_076874.8 NP_509275.2 4 - 5 446 119
NR_132546.1 3 - 4 727 119

View Full Product Details