Assay Design



Gene Transcripts

Gene Symbol
adt-2
Entrez Gene ID
181022
Gene Name
protein-coding
Gene Aliases
CELE_F08C6.1
Chromosome Location
Chr.X: 7586209 - 7595416 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029361.5 NP_001024532.2 2 - 3 118 136
NM_001375070.2 NP_001361970.1 1 - 1 118 136
GenBank mRNA AB072347.1 - 2 - 3 118 136

View Full Product Details