Assay Design
Gene Transcripts
- Gene Symbol
- dhs-29
- Entrez Gene ID
-
181027
- Gene Name
- DeHydrogenases, Short chain
- Gene Aliases
-
CELE_F27D9.6
- Chromosome Location
-
Chr.X: 7661891 - 7664919 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076893.4 |
NP_509294.1 |
6 - 7 |
672 |
100 |
View Full Product Details