Assay Design



Gene Transcripts

Gene Symbol
spp-14
Entrez Gene ID
181032
Gene Name
Saposin B-type domain-containing protein
Gene Aliases
CELE_K09F5.3
Chromosome Location
Chr.X: 7725214 - 7726390 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047806.6 NP_001041271.1 2 - 3 234 95

View Full Product Details