Assay Design
Gene Transcripts
- Gene Symbol
- hlh-8
- Entrez Gene ID
-
181069
- Gene Name
- Twist-related protein
- Gene Aliases
-
CELE_C02B8.4
- Chromosome Location
-
Chr.X: 8116457 - 8120340 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_076966.3 |
NP_509367.1 |
5 - 6 |
550 |
66 |
GenBank mRNA |
AF037063.1 |
- |
4 - 5 |
455 |
66 |
View Full Product Details