Assay Design
Gene Transcripts
- Gene Symbol
- spp-13
- Entrez Gene ID
-
184191
- Gene Name
- Saposin B-type domain-containing protein
- Gene Aliases
-
CELE_F08F1.6
- Chromosome Location
-
Chr.X: 8417419 - 8418758 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383615.1 |
|
2 - 3 |
91 |
100 |
View Full Product Details