Assay Design
Gene Transcripts
- Gene Symbol
- unc-27
- Entrez Gene ID
-
181124
- Gene Name
- Troponin I 2
- Gene Aliases
-
CELE_ZK721.2
- Chromosome Location
-
Chr.X: 8789450 - 8790621 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077087.8 |
NP_509488.2 |
1 - 2 |
159 |
105 |
View Full Product Details