Assay Design



Gene Transcripts

Gene Symbol
tsp-11
Entrez Gene ID
192064
Gene Name
protein-coding
Gene Aliases
CELE_B0563.2
Chromosome Location
Chr.X: 9163164 - 9166443 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001375094.1 NP_001362077.1 5 - 6 550 114
NM_077145.7 NP_509546.4 5 - 6 550 114

View Full Product Details