Assay Design



Gene Transcripts

Gene Symbol
pes-8
Entrez Gene ID
191742
Gene Name
protein-coding
Gene Aliases
CELE_F18G5.2
Chromosome Location
Chr.X: 9251788 - 9255778 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373414.1 NP_001360736.1 9 - 10 1229 73
NM_001373415.1 NP_001360737.1 8 - 9 1094 73

View Full Product Details