Assay Design
Gene Transcripts
- Gene Symbol
- fipr-21
- Entrez Gene ID
-
181238
- Gene Name
- FIP (Fungus-Induced Protein) Related
- Gene Aliases
-
CELE_F41E7.5
- Chromosome Location
-
Chr.X: 10304529 - 10304867 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077326.6 |
NP_509727.1 |
1 - 1 |
283 |
85 |
View Full Product Details