Assay Design
Gene Transcripts
- Gene Symbol
- elt-2
- Entrez Gene ID
-
181250
- Gene Name
- Transcription factor elt-2
- Gene Aliases
-
CELE_C33D3.1
- Chromosome Location
-
Chr.X: 10481240 - 10483446 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077354.5 |
NP_509755.2 |
4 - 5 |
512 |
78 |
View Full Product Details