Assay Design
Gene Transcripts
- Gene Symbol
- git-1
- Entrez Gene ID
-
181253
- Gene Name
- GIT1 (mammalian G protein-coupled receptor kinase InTeractor 1) homolog
- Gene Aliases
-
CELE_F14F3.2
- Chromosome Location
-
Chr.X: 10524898 - 10528469 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077360.8 |
NP_509761.2 |
7 - 8 |
968 |
58 |
View Full Product Details