Assay Design
Gene Transcripts
- Gene Symbol
- syd-2
- Entrez Gene ID
-
181255
- Gene Name
- Liprin-alpha
- Gene Aliases
-
CELE_F59F5.6
- Chromosome Location
-
Chr.X: 10549100 - 10555241 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077367.9 |
NP_509768.1 |
8 - 9 |
1103 |
111 |
GenBank mRNA |
AF170122.1 |
- |
7 - 8 |
1030 |
111 |
View Full Product Details