Assay Design



Gene Transcripts

Gene Symbol
daf-12
Entrez Gene ID
181263
Gene Name
protein-coding
Gene Aliases
CELE_F11A1.3
Chromosome Location
Chr.X: 10644440 - 10666875 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029376.6 NP_001024547.1 12 - 13 1460 93
NM_001047774.5 NP_001041239.1 11 - 12 1358 93
NM_001381088.1 NP_001366986.1 10 - 11 1283 93
GenBank mRNA AF136238.1 - 12 - 13 1493 93
AF136239.1 - 10 - 11 1290 93

View Full Product Details