Assay Design
Gene Transcripts
- Gene Symbol
- pgp-8
- Entrez Gene ID
-
181277
- Gene Name
- P-GlycoProtein related
- Gene Aliases
-
CELE_T21E8.3
- Chromosome Location
-
Chr.X: 10859301 - 10864422 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077410.6 |
NP_509811.2 |
9 - 10 |
2143 |
106 |
View Full Product Details