Assay Design
Gene Transcripts
- Gene Symbol
- pbo-4
- Entrez Gene ID
-
181285
- Gene Name
- Na(+)/H(+) exchanger protein 7
- Gene Aliases
-
CELE_K09C8.1
- Chromosome Location
-
Chr.X: 10954134 - 10958396 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077429.8 |
NP_509830.2 |
6 - 7 |
1011 |
120 |
GenBank mRNA |
AF497831.1 |
- |
6 - 7 |
1033 |
120 |
View Full Product Details