Assay Design
Gene Transcripts
- Gene Symbol
- lge-1
- Entrez Gene ID
-
187206
- Gene Name
- Glycosyltransferase-like protein LARGE
- Gene Aliases
-
CELE_K09C8.4
- Chromosome Location
-
Chr.X: 10966551 - 10969901 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077432.6 |
NP_509833.3 |
5 - 6 |
723 |
102 |
View Full Product Details