Assay Design
Gene Transcripts
- Gene Symbol
- hst-2
- Entrez Gene ID
-
181309
- Gene Name
- Heparan sulfate 2-O-sulfotransferase hst-2
- Gene Aliases
-
CELE_C34F6.4
- Chromosome Location
-
Chr.X: 11204103 - 11207155 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077470.6 |
NP_509871.2 |
1 - 2 |
207 |
87 |
GenBank mRNA |
AY552606.1 |
- |
1 - 2 |
140 |
87 |
View Full Product Details