Assay Design
Gene Transcripts
- Gene Symbol
- sto-4
- Entrez Gene ID
-
181350
- Gene Name
- Stomatin-4
- Gene Aliases
-
CELE_Y71H9A.3
- Chromosome Location
-
Chr.X: 11644126 - 11647425 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077543.5 |
NP_509944.1 |
6 - 7 |
733 |
71 |
View Full Product Details