Assay Design
Gene Transcripts
- Gene Symbol
- gst-42
- Entrez Gene ID
-
183911
- Gene Name
- Probable maleylacetoacetate isomerase
- Gene Aliases
-
CELE_D1053.1
- Chromosome Location
-
Chr.X: 11726264 - 11727936 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077561.8 |
NP_509962.1 |
2 - 3 |
270 |
75 |
View Full Product Details