Assay Design
Gene Transcripts
- Gene Symbol
- lsd-1
- Entrez Gene ID
-
181369
- Gene Name
- Probable lysine-specific histone demethylase 1
- Gene Aliases
-
CELE_T08D10.2
- Chromosome Location
-
Chr.X: 11947427 - 11951937 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077599.6 |
NP_510000.2 |
2 - 3 |
387 |
90 |
View Full Product Details