Assay Design
Gene Transcripts
- Gene Symbol
- lgc-23
- Entrez Gene ID
-
182622
- Gene Name
- Neur_chan_LBD domain-containing protein
- Gene Aliases
-
CELE_C15A7.1
- Chromosome Location
-
Chr.X: 12065208 - 12067433 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077615.8 |
NP_510016.2 |
1 - 2 |
197 |
87 |
View Full Product Details