Assay Design
Gene Transcripts
- Gene Symbol
- otpl-8
- Entrez Gene ID
-
186752
- Gene Name
- OToPetrin-Like
- Gene Aliases
-
CELE_H19J13.1
- Chromosome Location
-
Chr.X: 12266953 - 12270941 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077644.6 |
NP_510045.2 |
2 - 3 |
186 |
61 |
View Full Product Details