Assay Design
Gene Transcripts
- Gene Symbol
- srd-47
- Entrez Gene ID
-
184607
- Gene Name
- Serpentine receptor class delta-47
- Gene Aliases
-
CELE_F17A2.11
- Chromosome Location
-
Chr.X: 12333642 - 12335290 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077665.1 |
NP_510066.1 |
2 - 3 |
420 |
109 |
View Full Product Details