Assay Design
Gene Transcripts
- Gene Symbol
- zig-2
- Entrez Gene ID
-
192087
- Gene Name
- Zwei Ig domain protein zig-2
- Gene Aliases
-
CELE_F42F12.2
- Chromosome Location
-
Chr.X: 12340377 - 12341438 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077668.5 |
NP_510069.1 |
1 - 2 |
129 |
67 |
GenBank mRNA |
AF456249.1 |
- |
1 - 2 |
74 |
67 |
View Full Product Details