Assay Design
Gene Transcripts
- Gene Symbol
- myo-2
- Entrez Gene ID
-
181404
- Gene Name
- Myosin-2
- Gene Aliases
-
CELE_T18D3.4
- Chromosome Location
-
Chr.X: 12468061 - 12475348 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077691.5 |
NP_510092.2 |
10 - 11 |
5506 |
90 |
View Full Product Details