Assay Design



Gene Transcripts

Gene Symbol
tbc-13
Entrez Gene ID
181406
Gene Name
Rab-GAP TBC domain-containing protein
Gene Aliases
CELE_F45E6.3
Chromosome Location
Chr.X: 12485053 - 12490970 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373530.1 NP_001359553.1 8 - 9 1112 81

View Full Product Details