Assay Design
Gene Transcripts
- Gene Symbol
- skr-19
- Entrez Gene ID
-
181446
- Gene Name
- Skp1_POZ domain-containing protein
- Gene Aliases
-
CELE_R12H7.3
- Chromosome Location
-
Chr.X: 13224241 - 13224708 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF440518.1 |
- |
1 - 1 |
942 |
128 |
View Full Product Details