Assay Design
Gene Transcripts
- Gene Symbol
- cyp-14A2
- Entrez Gene ID
-
187184
- Gene Name
- CYtochrome P450 family
- Gene Aliases
-
CELE_K09A11.3
- Chromosome Location
-
Chr.X: 13267562 - 13269715 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077803.3 |
NP_510204.1 |
1 - 2 |
167 |
101 |
View Full Product Details