Assay Design



Gene Transcripts

Gene Symbol
srd-43
Entrez Gene ID
187577
Gene Name
Serpentine Receptor, class D (delta)
Gene Aliases
CELE_R04D3.9
Chromosome Location
Chr.X: 13303905 - 13305844 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322642.1 NP_001309448.1 4 - 5 697 79

View Full Product Details