Assay Design
Gene Transcripts
- Gene Symbol
- mxl-3
- Entrez Gene ID
-
181457
- Gene Name
- Protein mxl-3
- Gene Aliases
-
CELE_F46G10.6
- Chromosome Location
-
Chr.X: 13317965 - 13359942 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077822.6 |
NP_510223.1 |
4 - 5 |
625 |
71 |
View Full Product Details