Assay Design



Gene Transcripts

Gene Symbol
cah-4
Entrez Gene ID
181478
Gene Name
protein-coding
Gene Aliases
CELE_R01E6.3
Chromosome Location
Chr.X: 13542567 - 13544599 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_077863.8 NP_510264.1 5 - 6 677 98
NM_077864.5 NP_510265.1 5 - 6 807 98

View Full Product Details