Assay Design
Gene Transcripts
- Gene Symbol
- lgc-21
- Entrez Gene ID
-
181516
- Gene Name
- Neur_chan_LBD domain-containing protein
- Gene Aliases
-
CELE_C33G3.3
- Chromosome Location
-
Chr.X: 14064313 - 14067984 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077940.3 |
NP_510341.2 |
6 - 7 |
712 |
69 |
View Full Product Details