Assay Design
Gene Transcripts
- Gene Symbol
- C33G3.4
- Entrez Gene ID
-
181517
- Gene Name
- Probable beta-mannosidase
- Gene Aliases
-
CELE_C33G3.4
- Chromosome Location
-
Chr.X: 14071162 - 14076298 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077941.7 |
NP_510342.1 |
8 - 9 |
1594 |
147 |
View Full Product Details