Assay Design
Gene Transcripts
- Gene Symbol
- srx-54
- Entrez Gene ID
-
190845
- Gene Name
- G_PROTEIN_RECEP_F1_2 domain-containing protein
- Gene Aliases
-
CELE_Y102A5C.22
- Chromosome Location
-
Chr.V: 16975219 - 16976717 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074892.2 |
NP_507293.2 |
7 - 8 |
869 |
94 |
View Full Product Details