Assay Design
Gene Transcripts
- Gene Symbol
- srbc-62
- Entrez Gene ID
-
188596
- Gene Name
- Serpentine Receptor, class BC (class B-like)
- Gene Aliases
-
CELE_T19C9.1
- Chromosome Location
-
Chr.V: 17217990 - 17219004 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_074959.1 |
NP_507360.1 |
1 - 2 |
172 |
152 |
View Full Product Details