Assay Design
Gene Transcripts
- Gene Symbol
- lgc-16
- Entrez Gene ID
-
187975
- Gene Name
- Neur_chan_LBD domain-containing protein
- Gene Aliases
-
CELE_T01H10.5
- Chromosome Location
-
Chr.X: 12118640 - 12120661 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_077626.2 |
NP_510027.2 |
4 - 5 |
414 |
132 |
View Full Product Details